Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hba-a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008218.2 | NP_032244.2 | 2 | 166 | ||
GenBank mRNA | AK003077.1 | - | 2 | 164 | ||
AK010923.1 | - | 2 | 164 | |||
AK010927.1 | - | 2 | 165 | |||
AK010933.1 | - | 2 | 164 | |||
AK010937.1 | - | 2 | 167 | |||
AK010987.1 | - | 2 | 164 | |||
AK011018.1 | - | 2 | 166 | |||
AK011023.1 | - | 2 | 163 | |||
AK011025.1 | - | 2 | 166 | |||
AK011028.1 | - | 2 | 163 | |||
AK011055.1 | - | 2 | 167 | |||
AK011063.1 | - | 2 | 171 | |||
AK011076.1 | - | 2 | 164 | |||
AK011116.1 | - | 677 | ||||
AK012329.1 | - | 2 | 164 | |||
AK012755.1 | - | 2 | 164 | |||
AK171893.1 | - | 2 | 164 | |||
BC043020.1 | - | 2 | 146 | 5684314 |
Gene Symbol: Hba-a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083955.1 | NP_001077424.1 | 163 | |||
GenBank mRNA | AK005434.1 | - | 164 | |||
AK010422.1 | - | 164 | ||||
AK010938.1 | - | 165 | ||||
AK011030.1 | - | 164 | ||||
AK011073.1 | - | 164 | ||||
AK011121.1 | - | 164 | ||||
AK012348.1 | - | 164 | ||||
AK019138.1 | - | 165 | ||||
AK028173.1 | - | 164 | ||||
AK076068.1 | - | 164 | ||||
CF740193.1 | - | 150 | ||||
M10466.1 | - | 161 | ||||
M26895.1 | - | 137 |