Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008230.6 | NP_032256.3 | 3, 4 | 445 | ||
GenBank mRNA | AF109137.1 | - | 3, 4 | 355 | ||
AK076384.1 | - | 3, 4 | 429 | |||
AK088545.1 | - | 3, 4 | 432 | |||
AK133455.1 | - | 3, 4 | 446 | |||
AK150168.1 | - | 3, 4 | 433 | |||
AK153104.1 | - | 3, 4 | 433 | |||
BC052833.1 | - | 3, 4 | 430 | 30044098 | ||
X57437.1 | - | 3, 4 | 410 |