Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032572.1 | - | 10 | 2890 | ||
AK143616.1 | - | 14 | 1821 | |||
AK144669.1 | - | 14 | 1828 | |||
AK146813.1 | - | 14 | 1790 | |||
AK146918.1 | - | 14 | 1798 | |||
BC003741.1 | - | 14 | 1787 | 3488803 | ||
D63850.1 | - | 14 | 1804 | |||
FN687734.2 | - | 14 | 1547 |