Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010414.3 | NP_034544.1 | 33 | 4412 | ||
GenBank mRNA | L23312.1 | - | 33 | 4359 | ||
L23313.1 | - | 33 | 4359 | |||
L28827.1 | - | 33 | 4331 | |||
U24233.1 | - | 33 | 4372 |