Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hemt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010416.2 | NP_034546.2 | 3 | 300 | ||
GenBank mRNA | AK005659.1 | - | 3 | 305 | ||
AK005823.1 | - | 2 | 73 | |||
AK006826.1 | - | 3 | 304 |