Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019944.2 | NP_064328.2 | 3 | 1080 | ||
GenBank mRNA | AF153046.1 | - | 1080 | |||
BC132283.1 | - | 3 | 993 | 40130560 | ||
BC144718.1 | - | 3 | 994 |