Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010441.2 | NP_034571.1 | 2 | 1021 | ||
GenBank mRNA | AK028346.1 | - | 2 | 351 | ||
AK142059.1 | - | 2 | 383 | |||
AK147859.1 | - | 2 | 379 | |||
BC052158.1 | - | 2 | 555 | 6506864 | ||
BC069985.1 | - | 2 | 249 | 6332337 | ||
BC085085.1 | - | 2 | 322 | 6334495 | ||
X58380.1 | - | 2 | 235 |