Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010465.2 | NP_034595.2 | 1 | 265 | ||
GenBank mRNA | AB221685.1 | - | 1 | 221 | ||
AK050798.1 | - | 1 | 33 | |||
BC028482.1 | - | 1 | 265 | |||
BC141061.1 | - | 1 | 244 | 9055673 | ||
BX528590.1 | - | 1 | 266 | |||
X12504.1 | - | 2 | 290 | |||
X16510.1 | - | 226 | ||||
X16511.1 | - | 1 | 348 |