Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018951.3 | NP_061824.3 | 2 | 1211 | ||
NR_037939.1 | 2 | 839 | ||||
GenBank mRNA | AK312450.1 | - | 2 | 631 | ||
BC007600.1 | - | 3 | 856 | |||
BC013971.2 | - | 2 | 1179 | 4107013 | ||
BC071843.1 | - | 2 | 474 | |||
BC094807.1 | - | 2 | 474 | |||
M30599.1 | - | 1 | 230 |
Gene Symbol: HOXA10-HOXA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FY213345.1 | - | 2 | 352 |