Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010477.4 | NP_034607.3 | 5 | 748 | ||
GenBank mRNA | AK088844.1 | - | 5 | 662 | ||
AK143882.1 | - | 5 | 653 | |||
AK146831.1 | - | 5 | 700 | |||
AK177219.1 | - | 2 | 173 | |||
AK179525.1 | - | 2 | 172 | |||
AK188312.1 | - | 2 | 172 | |||
AK216069.1 | - | 2 | 142 | |||
BC016400.1 | - | 5 | 634 | 3585270 | ||
BC106112.1 | - | 5 | 649 | |||
X53584.1 | - | 4 | 538 | |||
X55023.1 | - | 5 | 610 |