Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008304.2 | NP_032330.1 | 2 | 616 | ||
GenBank mRNA | AK011042.1 | - | 2 | 465 | ||
AK204318.1 | - | 1 | 43 | |||
AK216657.1 | - | 1 | 43 | |||
BC047144.1 | - | 2 | 482 | 6395969 | ||
U00674.1 | - | 2 | 616 |