Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010493.2 | NP_034623.1 | 3 | 681 | ||
GenBank mRNA | AK149748.1 | - | 3 | 657 | ||
AK149781.1 | - | 3 | 656 | |||
AK149945.1 | - | 3 | 656 | |||
AK150003.1 | - | 3 | 657 | |||
AK150049.1 | - | 3 | 656 | |||
AK150057.1 | - | 3 | 657 | |||
AK150141.1 | - | 2 | 394 | |||
AK151227.1 | - | 3 | 657 | |||
AK151681.1 | - | 3 | 657 | |||
AK152155.1 | - | 3 | 657 | |||
AK152530.1 | - | 3 | 657 | |||
AK156417.1 | - | 3 | 657 | |||
AK168275.1 | - | 3 | 657 | |||
AK171502.1 | - | 3 | 657 | |||
AK171520.1 | - | 2 | 394 | |||
AK172321.1 | - | 3 | 657 | |||
BC008626.1 | - | 3 | 636 | 3588949 | ||
CT010246.1 | - | 3 | 606 | |||
CT010302.1 | - | 3 | 606 | |||
M31585.1 | - | 3 | 664 | |||
X16624.1 | - | 3 | 635 | |||
X52264.1 | - | 3 | 628 |