Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002147.3 | NP_002138.1 | 1 | 157 | ||
GenBank mRNA | AK314964.1 | - | 1 | 214 | ||
BC117247.1 | - | 1 | 118 | 40125798 | ||
BQ073272.1 | - | 91 | ||||
M92299.1 | - | 1 | 373 |