Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008331.3 | NP_032357.2 | 2 | 864 | ||
GenBank mRNA | AK089608.1 | - | 2 | 871 | ||
AK131669.1 | - | 2 | 862 | |||
AK144230.1 | - | 2 | 869 | |||
AK150352.1 | - | 2 | 871 | |||
AK150424.1 | - | 2 | 865 | |||
AK150609.1 | - | 2 | 874 | |||
AK151125.1 | - | 2 | 874 | |||
AK151158.1 | - | 2 | 865 | |||
AK151278.1 | - | 2 | 874 | |||
AK151298.1 | - | 2 | 868 | |||
AK151983.1 | - | 2 | 865 | |||
AK152112.1 | - | 2 | 867 | |||
AK152753.1 | - | 2 | 865 | |||
AK152853.1 | - | 1 | 539 | |||
AK153184.1 | - | 2 | 867 | |||
AK153500.1 | - | 2 | 865 | |||
AK153532.1 | - | 2 | 874 | |||
AK157848.1 | - | 2 | 865 | |||
BC003768.1 | - | 2 | 848 | 3496673 | ||
CT010226.1 | - | 1 | 770 | |||
U43084.1 | - | 2 | 839 |