Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206871.1 | NP_996754.1 | 1 | 87 | ||
GenBank mRNA | BC151087.1 | - | 1 | 227 | 9088000 |
Gene Symbol: Ifna12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177361.2 | NP_796335.1 | 1 | 155 | ||
GenBank mRNA | AY190046.1 | - | 1 | 155 | ||
BC104352.1 | - | 1 | 134 | 40049569 | ||
BC104353.1 | - | 1 | 134 | 40049571 |
Gene Symbol: Ifna13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177347.2 | NP_796321.1 | 1 | 161 | ||
GenBank mRNA | AY190047.1 | - | 1 | 161 | ||
BC120724.1 | - | 1 | 117 | 40129647 | ||
BC120750.1 | - | 1 | 117 | 40129673 |
Gene Symbol: Ifna14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206975.1 | NP_996858.1 | 1 | 87 | ||
GenBank mRNA | BC120722.1 | - | 1 | 137 | 40129645 | |
BC125321.1 | - | 1 | 137 | 40129836 |
Gene Symbol: Ifna15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206870.1 | NP_996753.1 | 1 | 87 | ||
GenBank mRNA | BC116216.2 | - | 1 | 87 | 40091189 |
Gene Symbol: Ifnab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008336.2 | NP_032362.2 | 1 | 128 | ||
GenBank mRNA | BC116862.1 | - | 1 | 128 | 40126181 | |
BC116864.1 | - | 1 | 128 | 40126183 |