Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igtp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018738.4 | NP_061208.3 | 3 | 1621 | ||
GenBank mRNA | AK002843.1 | - | 3 | 1559 | ||
AK088315.1 | - | 3 | 1620 | |||
AK149817.1 | - | 3 | 1550 | |||
AK151779.1 | - | 3 | 1561 | |||
AK151860.1 | - | 3 | 1542 | |||
AK152337.1 | - | 4 | 1393 | |||
AK152483.1 | - | 3 | 1542 | |||
AK153436.1 | - | 3 | 1561 | |||
AK165065.1 | - | 4 | 1449 | |||
AK169307.1 | - | 3 | 1580 | |||
AY225483.1 | - | 2 | 1186 | |||
BC132557.1 | - | 2 | 1249 | 40130834 | ||
BC138607.1 | - | 2 | 1249 | 8861627 | ||
BC145346.1 | - | 3 | 1050 | |||
U53219.1 | - | 3 | 1533 |