Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igtp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018738.4 | NP_061208.3 | 3 | 677 | ||
GenBank mRNA | AK002843.1 | - | 3 | 615 | ||
AK088315.1 | - | 3 | 676 | |||
AK149817.1 | - | 3 | 606 | |||
AK151779.1 | - | 3 | 617 | |||
AK151860.1 | - | 3 | 598 | |||
AK152483.1 | - | 3 | 598 | |||
AK153436.1 | - | 3 | 617 | |||
AK169307.1 | - | 3 | 636 | |||
AY225483.1 | - | 2 | 242 | |||
BC132557.1 | - | 2 | 305 | 40130834 | ||
BC138607.1 | - | 2 | 305 | 8861627 | ||
U53219.1 | - | 3 | 589 |