Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159774.1 | NP_001153246.1 | 9 | 863 | ||
NM_010546.2 | NP_034676.1 | 9 | 863 | |||
GenBank mRNA | AF026524.1 | - | 8 | 731 | ||
AF088910.1 | - | 9 | 923 | |||
AK086442.1 | - | 7 | 680 | |||
AK144603.1 | - | 9 | 856 | |||
AK150884.1 | - | 9 | 815 | |||
AK151888.1 | - | 9 | 815 | |||
AK151955.1 | - | 9 | 815 | |||
AK153151.1 | - | 9 | 815 | |||
AK155587.1 | - | 9 | 796 | |||
AK156294.1 | - | 9 | 821 | |||
BC004772.1 | - | 9 | 801 | |||
BC037723.1 | - | 9 | 835 | 4482634 |