Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il11ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163401.1 | NP_001156873.1 | 7, 8 | 755 | ||
NM_001172054.1 | NP_001165525.1 | 8, 9 | 841 | |||
NM_010549.3 | NP_034679.1 | 7, 8 | 760 | |||
GenBank mRNA | AK146742.1 | - | 7, 8 | 708 | ||
AK159559.1 | - | 7, 8 | 690 | |||
BC004619.1 | - | 7, 8 | 679 | 3585497 | ||
U14412.1 | - | 7, 8 | 690 | |||
X74953.1 | - | 7, 8 | 713 |