Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Il18rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010553.4 | NP_034683.1 | 4, 5 | 490 | ||
GenBank mRNA | AF077347.1 | - | 4, 5 | 499 | ||
AK053176.1 | - | 4, 5 | 405 | |||
AK137541.1 | - | 4, 5 | 451 | |||
BC120598.1 | - | 4, 5 | 497 | 40129521 | ||
BC120600.1 | - | 4, 5 | 497 | 40129523 |