Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159317.1 | NP_001152789.1 | 8 | 1163 | ||
NM_001159318.1 | NP_001152790.1 | 8 | 1163 | |||
NM_008364.2 | NP_032390.1 | 8 | 1163 | |||
NM_134103.2 | NP_598864.1 | 8 | 1163 | |||
GenBank mRNA | AK033972.1 | - | 8 | 1112 | ||
AK034269.1 | - | 8 | 1127 | |||
AK039582.1 | - | 8 | 1115 | |||
AK045686.1 | - | 8 | 1112 | |||
AK083209.1 | - | 8 | 1160 | |||
AK136782.1 | - | 8 | 1114 | |||
AK149467.1 | - | 8 | 1112 | |||
BC021159.1 | - | 8 | 1112 | 4161899 | ||
X85999.1 | - | 8 | 1114 |