Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143669.1 | - | 7 | 932 | ||
AK153873.1 | - | 7 | 947 | |||
BC014720.1 | - | 7 | 926 | 4924951 | ||
D13565.1 | - | 7 | 924 | |||
D13821.1 | - | 7 | 919 | |||
KC815469.1 | - | 7 | 818 | |||
L20048.1 | - | 7 | 936 | |||
X75337.1 | - | 7 | 949 |