Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008377.2 | NP_032403.2 | 18 | 3354 | ||
GenBank mRNA | AK133771.1 | - | 5 | 922 | ||
BC141300.1 | - | 18 | 3146 | 9055912 | ||
BC145623.1 | - | 17 | 2999 | 9054191 | ||
CN699905.1 | - | 2 | 229 | |||
D78572.1 | - | 18 | 3347 |