Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010564.4 | NP_034694.3 | 2 | 1231 | ||
GenBank mRNA | BC056627.1 | - | 2 | 1231 | 5009577 | |
X55957.1 | - | 2 | 917 | |||
X69618.1 | - | 2 | 1083 |