Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008392.1 | NP_032418.1 | 5 | 1513 | ||
GenBank mRNA | AK036446.1 | - | 5 | 1549 | ||
AK089891.1 | - | 5 | 1497 | |||
AK152177.1 | - | 5 | 1512 | |||
AK152248.1 | - | 5 | 1512 | |||
AK152332.1 | - | 5 | 1512 | |||
AK152635.1 | - | 5 | 1512 | |||
AK153013.1 | - | 5 | 1512 | |||
AK172001.1 | - | 5 | 1512 | |||
AK172298.1 | - | 5 | 1512 | |||
L38281.1 | - | 5 | 1729 |