Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010576.3 | NP_034706.3 | 25, 26 | 3028 | ||
GenBank mRNA | AB850880.1 | - | 25, 26 | 2793 | ||
AF109136.1 | - | 25, 26 | 2864 | |||
BC068313.1 | - | 25, 26 | 2808 | 30093248 | ||
X53176.1 | - | 25, 26 | 2951 |