Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010576.3 | NP_034706.3 | 21 | 2529 | ||
GenBank mRNA | AB850880.1 | - | 21 | 2294 | ||
AF109136.1 | - | 21 | 2365 | |||
X53176.1 | - | 21 | 2452 |