Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008399.2 | NP_032425.2 | 17 | 2053 | ||
NM_172944.2 | NP_766532.1 | 17 | 2053 | |||
GenBank mRNA | AK040983.1 | - | 17 | 2054 | ||
AK154021.1 | - | 17 | 2051 | |||
BC150690.1 | - | 17 | 2053 | 9087601 | ||
DQ386608.1 | - | 17 | 2053 | |||
U12236.1 | - | 17 | 2077 |