Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008407.2 | NP_032433.2 | 19 | 2112 | ||
GenBank mRNA | AK146700.1 | - | 18 | 2601 | ||
BC015276.1 | - | 19 | 2070 | 4212290 | ||
X70393.1 | - | 18 | 2069 |