Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008408.4 | NP_032434.3 | 15, 16 | 1835 | ||
GenBank mRNA | AK030363.1 | - | 15, 16 | 1899 | ||
AK050050.1 | - | 1, 2 | 103 | |||
AK077877.1 | - | 15, 16 | 1858 | |||
AK081547.1 | - | 15, 16 | 1892 | |||
AK087470.1 | - | 15, 16 | 1882 | |||
AK129027.1 | - | 15, 16 | 1873 | |||
AK150640.1 | - | 15, 16 | 1880 | |||
AK151541.1 | - | 15, 16 | 1891 | |||
AK151791.1 | - | 15, 16 | 1891 | |||
AK152242.1 | - | 15, 16 | 1880 | |||
AK153840.1 | - | 15, 16 | 1836 | |||
AK153886.1 | - | 15, 16 | 1936 | |||
AK160037.1 | - | 15, 16 | 1874 | |||
AK160607.1 | - | 15, 16 | 1836 | |||
AK168505.1 | - | 15, 16 | 1881 | |||
AK168937.1 | - | 15, 16 | 1880 | |||
AK169669.1 | - | 15, 16 | 1859 | |||
AK216214.1 | - | 3, 4 | 232 | |||
AX799090.1 | - | 15, 16 | 1878 | |||
BC028897.1 | - | 14, 15 | 1784 | |||
BC037612.1 | - | 15, 16 | 1874 | 5353797 | ||
BC085313.1 | - | 15, 16 | 1864 | 30443880 | ||
L34260.1 | - | 15, 16 | 1878 |