Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048177.2 | NP_001041642.1 | 22, 23 | 3521 | ||
NM_008413.3 | NP_032439.2 | 23, 24 | 3604 | |||
GenBank mRNA | BC054807.1 | - | 22, 23 | 3284 | 6838318 | |
BC059834.1 | - | 22, 23 | 3284 | 6838318 | ||
L16956.1 | - | 22, 23 | 3273 |