Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008428.4 | NP_032454.1 | 3 | 993 | ||
GenBank mRNA | AK031777.1 | - | 3 | 975 | ||
AK156346.1 | - | 3 | 943 | |||
AK157219.1 | - | 2 | 537 | |||
BC003756.1 | - | 3 | 1030 | 3490123 | ||
D88159.1 | - | 3 | 949 |