Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008445.2 | NP_032471.2 | 7 | 3035 | ||
GenBank mRNA | AF013116.1 | - | 7 | 2611 | ||
AK029381.1 | - | 1 | 435 | |||
AK139894.1 | - | 7 | 711 | |||
AK147572.1 | - | 7 | 2837 | |||
AK200456.1 | - | 1 | 25 | |||
AK220283.1 | - | 8 | 2760 | |||
BC052732.1 | - | 7 | 2192 | |||
BC053053.1 | - | 7 | 2977 | |||
BC062648.1 | - | 7 | 2710 | |||
BC127063.1 | - | 7 | 2247 | 40109965 |