Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Kifap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133957.1 | - | 18 | 2124 | ||
AK179475.1 | - | 2 | 160 | |||
BC040362.1 | - | 18 | 2185 | 4506177 | ||
BC049100.1 | - | 18 | 2383 | 5698182 | ||
D50366.1 | - | 18 | 2298 | |||
D50367.1 | - | 18 | 2298 |