Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136069.2 | NP_001129541.2 | 4, 5 | 755 | ||
NM_010699.2 | NP_034829.1 | 4, 5 | 562 | |||
GenBank mRNA | AK089929.1 | - | 4, 5 | 526 | ||
AK146121.1 | - | 4, 5 | 523 | |||
AK149919.1 | - | 4, 5 | 523 | |||
AK150156.1 | - | 4, 5 | 526 | |||
AK167789.1 | - | 4, 5 | 524 | |||
AK167944.1 | - | 4, 5 | 524 | |||
AK168346.1 | - | 4, 5 | 523 | |||
AK168920.1 | - | 4, 5 | 528 | |||
AK170386.1 | - | 4, 5 | 527 | |||
AK170708.1 | - | 4, 5 | 524 | |||
AK171373.1 | - | 4, 5 | 525 | |||
AK201923.1 | - | 1, 2 | 86 | |||
AK207627.1 | - | 1, 2 | 86 | |||
AK209803.1 | - | 1, 2 | 86 | |||
AK213457.1 | - | 1, 2 | 86 | |||
AK214241.1 | - | 1, 2 | 86 | |||
AK214906.1 | - | 1, 2 | 86 | |||
AK215061.1 | - | 1, 2 | 86 | |||
AK219462.1 | - | 1, 2 | 86 | |||
BC005509.1 | - | 3, 4 | 374 | |||
BC094019.1 | - | 4, 5 | 479 | 4195220 | ||
BC094428.1 | - | 4, 5 | 516 | 30700835 | ||
U13687.1 | - | 4, 5 | 622 |