Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145953.1 | NP_001139425.1 | 4 | 584 | ||
NM_010705.3 | NP_034835.1 | 4 | 557 | |||
GenBank mRNA | AK008593.1 | - | 3 | 226 | ||
AK089249.1 | - | 4 | 501 | |||
AK149783.1 | - | 4 | 531 | |||
AK155244.1 | - | 4, 5 | 403 | |||
AK155484.1 | - | 4 | 499 | |||
AK159536.1 | - | 4 | 525 | |||
AK170803.1 | - | 4 | 614 | |||
BC132328.1 | - | 4 | 510 | 40130605 | ||
BC138790.1 | - | 4 | 510 | 8861812 | ||
BC145419.1 | - | 4 | 510 | 9053977 | ||
J03723.1 | - | 2 | 457 | |||
M33215.1 | - | 2 | 166 | |||
X16074.1 | - | 4 | 604 | |||
X16834.1 | - | 4 | 503 |