Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010706.2 | NP_034836.1 | 6, 7 | 939 | ||
GenBank mRNA | AF026795.1 | - | 5, 6 | 489 | ||
AF510729.1 | - | 6, 7 | 588 | |||
AK145734.1 | - | 6, 7 | 759 | |||
AY044870.1 | - | 6, 7 | 611 | |||
BC011236.1 | - | 6, 7 | 577 | 4205669 | ||
BC021632.1 | - | 6, 7 | 595 | 4218531 | ||
BC030297.1 | - | 6, 7 | 752 | 4986324 | ||
BC094008.1 | - | 6, 7 | 590 | 4986911 | ||
BC141106.1 | - | 6, 7 | 661 | 9055718 | ||
BC145387.1 | - | 6, 7 | 661 | 9053945 | ||
EF017938.1 | - | 6, 7 | 939 | |||
EF017939.1 | - | 6, 7 | 939 |