Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010730.2 | NP_034860.2 | 4 | 319 | ||
GenBank mRNA | AK002681.1 | - | 4 | 321 | ||
AK140838.1 | - | 4 | 322 | |||
AK150564.1 | - | 4 | 321 | |||
AK150718.1 | - | 4 | 321 | |||
AK153156.1 | - | 4 | 321 | |||
AK153490.1 | - | 4 | 321 | |||
AK167612.1 | - | 4 | 321 | |||
BC002289.1 | - | 3 | 274 | 3590168 | ||
BC004594.1 | - | 3 | 260 | 3587973 | ||
CT009771.1 | - | 3 | 245 | |||
CT010298.1 | - | 3 | 245 | |||
EU684130.1 | - | 3 | 245 | |||
X07486.1 | - | 3 | 258 |