Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010731.3 | NP_034861.3 | 2 | 550 | ||
GenBank mRNA | AF086830.1 | - | 2 | 167 | ||
AK149961.1 | - | 2 | 227 | |||
AK154907.1 | - | 2 | 244 | |||
AK157664.1 | - | 244 | ||||
BC024815.2 | - | 2 | 550 | |||
BC057204.1 | - | 2 | 188 | |||
BC138524.1 | - | 2 | 309 | 8861544 | ||
BC145311.1 | - | 2 | 349 | 9053866 | ||
BC171957.1 | - | 2 | 309 | 9054243 |