Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013587.2 | NP_038615.2 | 5 | 712 | ||
GenBank mRNA | AJ639614.1 | - | 5 | 719 | ||
AK089250.1 | - | 5 | 715 | |||
AK145710.1 | - | 5 | 755 | |||
AK150451.1 | - | 5 | 726 | |||
AK152485.1 | - | 5 | 726 | |||
AK153323.1 | - | 5 | 726 | |||
AK166616.1 | - | 5 | 715 | |||
AK166746.1 | - | 5 | 726 | |||
AK166819.1 | - | 5 | 730 | |||
AK169132.1 | - | 5 | 720 | |||
AK171471.1 | - | 5 | 715 | |||
AK193187.1 | - | 2 | 140 | |||
AK206813.1 | - | 3 | 248 | |||
BC032170.1 | - | 5 | 715 | |||
BC046641.1 | - | 5 | 704 | |||
BC057979.1 | - | 5 | 683 | |||
BC059887.1 | - | 5 | 529 | |||
BC094324.1 | - | 5 | 712 | 6402163 | ||
D00622.1 | - | 5 | 705 | |||
S67967.1 | - | 5 | 738 |