Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008528.4 | NP_032554.2 | 17 | 1802 | ||
GenBank mRNA | AB015290.1 | - | 17 | 1476 | ||
AF068182.1 | - | 17 | 1457 | |||
AK089704.1 | - | 17 | 1442 | |||
BC059785.1 | - | 17 | 1428 | 5339723 | ||
Y17159.1 | - | 17 | 1802 |