Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ly9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008534.3 | NP_032560.2 | 2 | 357 | ||
GenBank mRNA | AF244131.1 | - | 2 | 357 | ||
AK040306.1 | - | 2 | 308 | |||
AK042288.1 | - | 2 | 296 | |||
AK087345.1 | - | 2 | 277 | |||
AK134106.1 | - | 2 | 374 | |||
AK150121.1 | - | 2 | 284 | |||
BC055380.1 | - | 2 | 257 | |||
BC066212.1 | - | 2 | 245 | |||
M84412.1 | - | 2 | 221 |