Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010750.3 | NP_034880.1 | 1 | 1458 | ||
GenBank mRNA | AF040945.1 | - | 1 | 1445 | ||
AF228913.1 | - | 1 | 1445 | |||
AK032114.1 | - | 1 | 1429 | |||
AK043552.1 | - | 1 | 1458 | |||
BC014750.1 | - | 1 | 1407 | 4526962 |