Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010757.2 | NP_034887.1 | 3 | 847 | ||
GenBank mRNA | AK143654.1 | - | 3 | 853 | ||
AK143717.1 | - | 3 | 851 | |||
AK153856.1 | - | 3 | 817 | |||
BC014295.1 | - | 3 | 825 | 4189276 | ||
D42124.1 | - | 3 | 850 |