Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010759.1 | NP_034889.1 | 1 | 498 |
Gene Symbol: Mageb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC145851.1 | - | 2 | 443 | 40131154 |
Gene Symbol: Mageb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008545.2 | NP_032571.2 | 1 | 1221 | ||
GenBank mRNA | AK019658.1 | - | 1 | 1221 | ||
AY099463.1 | - | 1 | 489 | |||
AY099464.1 | - | 1 | 489 |