Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021516.4 | NP_067491.2 | 15 | 2351 | ||
NM_022801.4 | NP_073712.2 | 15 | 2351 | |||
GenBank mRNA | AF240782.1 | - | 15 | 1896 | ||
AF240783.1 | - | 15 | 1896 | |||
AK002654.1 | - | 6 | 913 | |||
AK075742.1 | - | 6 | 913 | |||
AK132247.1 | - | 15 | 2335 | |||
AK166157.1 | - | 15 | 2340 | |||
BC026445.1 | - | 8 | 1256 | 4459439 | ||
BC066071.1 | - | 14 | 2986 |