Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021516.4 | NP_067491.2 | 14 | 2059 | ||
NM_022801.4 | NP_073712.2 | 14 | 2059 | |||
GenBank mRNA | AF240782.1 | - | 14 | 1604 | ||
AF240783.1 | - | 14 | 1604 | |||
AK002654.1 | - | 5 | 621 | |||
AK075742.1 | - | 5 | 621 | |||
AK132247.1 | - | 14 | 2043 | |||
AK166157.1 | - | 14 | 2048 | |||
BC026445.1 | - | 7 | 964 | 4459439 | ||
BC066071.1 | - | 13 | 2694 | |||
CA575239.1 | - | 4 | 450 |