Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010771.6 | NP_034901.2 | 12 | 2336 | ||
GenBank mRNA | AK029510.1 | - | 11 | 1714 | ||
AK087939.1 | - | 12 | 2221 | |||
AK088351.1 | - | 12 | 2229 | |||
AK129199.1 | - | 12 | 2287 | |||
AK135937.1 | - | 11 | 2069 | |||
AK148384.1 | - | 5 | 663 | |||
AK157744.1 | - | 11 | 1432 | |||
AK161128.1 | - | 12 | 2235 | |||
BC029070.1 | - | 12 | 2247 | 3989914 | ||
BC090833.1 | - | 11 | 2109 | 6854095 | ||
CT010322.1 | - | 11 | 2020 |