Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL360193.1 | - | 1 | 391 | ||
BC057397.1 | - | 1 | 2249 | 6188453 | ||
BC063507.1 | - | 1 | 2255 | 4153196 |