Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mep1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008585.2 | NP_032611.2 | 7 | 446 | ||
GenBank mRNA | AK093914.1 | - | 7 | 407 | ||
AK143906.1 | - | 7 | 404 | |||
AK164734.1 | - | 7 | 401 | |||
BC015258.1 | - | 7 | 446 | 4241252 | ||
M74897.1 | - | 7 | 445 |